Learn More
anti-Plasminogen, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NB11068189
Catalog No. NB11068189
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Plasminogen Polyclonal antibody specifically detects Plasminogen in Chicken samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Plasminogen
8.5 mg/mL
Western Blot 1-5 ug/ml, ELISA 1:10000
Purified
PLG
Chicken Plasminogen
Protein A purified
RUO
Store at -80C. Avoid freeze-thaw cycles.
Polyclonal
Chicken Plasminogen and plasmin.
Western Blot, ELISA
Unconjugated
50mM Sodium Phosphate (pH 6.6), 0.1M NaCl and 1mM EDTA with No Preservative
DKFZp779M0222, EC 3.4.21, EC 3.4.21.7, plasmin, plasminogen
Rabbit
IgG
0.1 mg
Cell Cycle and Replication
Primary
5340
Chicken