Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLLP Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1086

Catalog No. PA1-086


Add to cart

Description

Description

PA1-086 detects plasmolipin from rat and human samples. PA1-086 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~18 kDa protein representing plasmolipin from rat kidney tissue extract. The PA1-086 immunogen is a synthetic peptides corresponding to residues C D(127) L T S L R G (S/T) R P Y N Q R S(141) of human and rat Plasmolipin. This peptide (Cat. No. PEP-250) can be used for neutralization and control experiments.

Plasmolipin is an 18 kDa membrane-bound proteolipid that belongs to the tetraspan (4TM) family, a rapidly growing group of myelin-associated proteins. Many of these proteins could be linked to human hereditary demyelinating neuropathies. Studies indicate that plasmolipin is a component of the cytoskeletal structure and has been localized to the apical surface of tubular epithelial cells. It has been shown that, upon addition to lipid bilayers, plasmolipin induces formation of ion channels that are both voltage-dependent and potassium-specific. Plasmolipin was originally isolated from kidney tissue. Subsequently, it has been shown to be widely expressed, not only being present in kidney, but also in the nervous system, thymus, testis, lung, and thyroid gland.

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

PLLP
Polyclonal
Unconjugated
PLLP
Liquid
PLLP
Synthetic peptides corresponding to residues C D(127) L T S L R G (S/T) R P Y N Q R S(141) of human and rat Plasmolipin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q9Y342, P47987
PLLP, Plasma membrane proteolipid
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
51090, 64364
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit