Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-PLEKHA3, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP183924
Description
PLEKHA3 Polyclonal antibody specifically detects PLEKHA3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PLEKHA3 | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FAPP-1, FAPP1PH domain-containing family A member 3, FLJ20067, four-phosphate-adaptor protein 1, Phosphatidylinositol-four-phosphate adapter protein 1, Phosphoinositol 4-phosphate adapter protein 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 3, pleckstrin homology domain-containing family A member 3, pleckstrin homology domain-containing, family A (phosphoinositide bindingspecific) member 3 | |
Rabbit | |
IgG | |
0.1 ml | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
Affinity Purified | |
PLEKHA3 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:QQVHTIQEFVHHDENHSSPSAENMNEASSLLSATCNTFITTLEECVKIANAKFKPEMFQLHHPDPLVSPVSPSPVQMMKRSVSHP | |
Immunogen affinity purified | |
RUO | |
Primary | |
65977 | |
Human, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only