Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pleckstrin homology domain containing, family G (with RhoGef domain) member 2, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human PLEKHG2 protein.

Manufacturer:  Abnova Corporation H00064857B01

Catalog No. 89-008-111


Add to cart

Description

Description

Sequence: MFPQNAKPGFKHAGSEGELYPPESQPPVSGSAPPEDLEDAGPPTLDPSGTSITEEILELLNQRGLRDPGPSTHDIPKFPGDSQVPGDSETLTFQALPSRDSSEEEEEEEEGLEMDERGPSPLHVLEGLESSIAAEMPSIPCLTKIPDVPNLPEIPSHCEIPEGSRLPSLSDISDVFEMPCLPAIPSVPNTPSLSSTPTLSCDSWLQGPLQEPAEAPATRRELFSGSNPGKLGEPPSGGKAGPEEDEEGVSFTDFQPQDVTQHQGFPDELAFRSCSEIRSAWQALEQGQLARPGFPEPLLILEDSDLGGDSGSGKAGAPSSERTASRVRELARLYSERIQQMQRAETRASANAPRRRPRVLAQPQPSPCLPQEQAEPGLLPAFGHVLVCELAFPLTCAQESVPLGPAVWVQAAIPLSKQGGSPDGQGLHVSNLPKQDLPGIHVSAATLLPEQGGSRHVQAPAATPLPKQEGPLHLQVPALTTFSDQGHPEIQVPATTPLPEHKSHMVIPAPSTAFCPEQGHCADIHVPTTPALPKEICSDFTVSVTTPVPKQEGHLDSESPTNIPLTKQGGSRDVQGPDPVCSQPIQPLSWHGSSLDPQGPGDTLPPLPCHLPDLQIPGTSPLPAHGSHLDHRIPANAPLSLSQELPDTQVPATTPLPLPQVLTDIWVQALPTSPKQGSLPDIQGPAAAPPLPEPSLTDTQVQKLTPSLEQKSLIDAHVPAATPLPERGGSLDIQGLSPTPVQTTMVLSKPGGSLASHVARLESSDLTPPHSPPPSSRQLLGPNAAALSRYLAASYISQSLARRQGPGGGAPAASRGSWSSAPTSRASSPPPQPQPPPPAARRLSYATTVNIHVGGGGRLRPAKAQVRLNHPALLASTQESMGLHRAQGAPDAPFHM
Specifications

Specifications

pleckstrin homology domain containing, family G (with RhoGef domain) member 2
Unconjugated
No additive
BC013426.1
CLG|DKFZp667J2325|FLJ00018|FLJ22458|FLJ38638
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
64857
Western Blot
Mouse polyclonal antibody raised against a full-length human PLEKHG2 protein.
PLEKHG2
Serum
PLEKHG2
PLEKHG2 (AAH13426.1, 1 a.a. ~ 896 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit