Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLXNA4 Mouse anti-Human, Clone: 707206, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524326

Catalog No. PIMA524326


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with Recombinant human Plexin A1, Recombinant mouse (rm) Plexin A1 or rmPlexin A2 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

Forms a receptor complex with neuropilin-1 and can propagate semaphorin-3A elicited inhibitory signals into cells and neurons.
Specifications

Specifications

PLXNA4
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q9HCM2
FAYV2820, PLEXA4, PLXNA4A, PLXNA4B, PRO34003, plexin-A4
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
91584
Flow Cytometry
707206
Unconjugated
PLXNA4
Lyophilized
PLXNA4
CHO-derived recombit human Plexin A4 Thr24-Pro1237
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit