Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PLK4, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133402

Catalog No. NBP133402


Add to cart

Description

Description

PLK4 Polyclonal antibody specifically detects PLK4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

PLK4
0.66mg/mL
Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
PLK4
Recombinant protein encompassing a sequence within the C-terminus region of human PLK4. The exact sequence is proprietary.
109 kDa
0.1 ml
Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers
Primary
10733
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300
EC 2.7.11.21, PLK-4, polo-like kinase 4 (Drosophila), polo-like kinase 4SAKSTK18serine/threonine kinase 18, Sak, serine/threonine protein kinase SAK, Serine/threonine-protein kinase 18, serine/threonine-protein kinase PLK4, Serine/threonine-protein kinase Sak, Snk akin kinase
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only