Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

PLOD2 Mouse anti-Human, Clone: 408105, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524145

Catalog No. PIMA524145



Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

PLOD2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
O00469
LH2, PLOD2, TLH, 2-oxoglutarate 5-dioxygenase (lysine hydroxylase) 2, 2-oxoglutarate 5-dioxygenase 2, procollagen-lysine, telopeptide lysyl hydroxylase
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5352
Immunoprecipitation, Western Blot
408105
Unconjugated
PLOD2
Lyophilized
PLOD2
Sf 21-derived recombit human Lysine Hydroxylase 2/PLOD2 Ala26-Pro758
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit