Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLTP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514601

Catalog No. PIPA514601

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PLTP is one of at least two lipid transfer proteins found in human plasma. The protein transfers phospholipids from triglyceride-rich lipoproteins to high density lipoprotein (HDL). In addition to regulating the size of HDL particles, this protein may be involved in cholesterol metabolism.
Specifications

Specifications

PLTP
Polyclonal
PBS with 0.09% sodium azide
P55058
Phospholipid transfer protein; Lipid transfer protein II; PLTP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5360
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PLTP
Liquid
PLTP
KLH conjugated synthetic peptide between 451-479 amino acids from the C-terminal region of human PLTP
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit