Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLZF Mouse anti-Human, APC, Clone: 6318100, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523681

Catalog No. PIMA523681


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with recombit human (rh) ZBTB38, rhZNF24, rhZNF143, rhZNF206, rhZNF281, or rhZNF423 is observed.

This gene is a member of the Krueppel C2H2-type zinc-finger protein family and encodes a zinc finger transcription factor that contains nine Kruppel-type zinc finger domains at the carboxyl terminus. This protein is located in the nucleus, is involved in cell cycle progression, and interacts with a histone deacetylase. Specific instances of aberrant gene rearrangement at this locus have been associated with acute promyelocytic leukemia (APL). Alternate transcriptional splice variants have been characterized.
Specifications

Specifications

PLZF
Monoclonal
APC
ZBTB16
Liquid
ZBTB16
E. coli-derived recombit human PLZF Met1-Gln254
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
6318100
Buffered saline solution with BSA and 0.09% sodium azide
Q05516
PLZF, ZNF145, zinc finger and BTB domain-containing protein 16, zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia), zinc finger protein PLZF
Mouse
IgG2A
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
7704
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit