Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMCH, Polyclonal, Invitrogen™

Catalog No. PIPA124289
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA124289 100 μg
1 options

Catalog No. PIPA124289

Supplier: Thermo Scientific PA124289

Rabbit Polyclonal Antibody

MCH may act as a neurotransmitter or neuromodulator in a broad array of neuronal functions directed toward the regulation of goal-directed behavior, such as food intake, and general arousal. May also have a role in spermatocyte differentiation.

PA1-24289 detects PMCH in Human samples.PA1-24289 has been successfully used in Western Blot procedures.PA1-24289 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human PMCH.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMCH
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene PMCH
Gene Accession No. P20382
Gene Alias MCH
Gene Symbols PMCH
Host Species Rabbit
Immunogen Synthetic peptide region of human PMCH.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5367
Target Species Human
Content And Storage Store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.