Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PML, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138472

Catalog No. 01-673-865

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The PML protein is a zinc finger transcription factor expressed as three major transcription products due to alternative splicing. The gene encoding human PML maps to chromosome 15q22. The t(15;17)(q22;q11.2-q12) chromosomal trans-location of the retinoic acid receptor alpha (RARalpha) gene occurs in virtually all cases of acute promyelocytic leukemia and results in the expression of a PML/RARalpha chimeric protein of 106kDa. Myeloid precursor cells expression the PML/RARalpha chimera fail to differentiate and exhibit an increased growth rate consequent to diminished apoptosis.

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is lymph node tissue.
Specifications

Specifications

PML
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P29590
promyelocytic leukemia protein
Rabbit
IgG
1mL
4° C, do not freeze
Primary
5371
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
PML
Liquid
PML
Synthetic peptide derived from near N-terminus of human PML
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit