Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMP22 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138475

Catalog No. 01-673-868

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is pancreas tissue.

PMP-22 (Peripheral Myelin Protein) is a glycoprotein expressed in the compact myolin of the peripheral nervous system. In the peripheral nervous system, PMP-22 is produced by myelinating Schwann cells and is coexpressed with the genes for myelin basic protein (MBP) and protein P0 during nerve development and regeneration. Alterations in the level of this protein cause several genetic human diseases. If the protein is duplicated, patients develop Charcot-Marie-Tooth disease. If one copy of the gene is deleted, they suffer from the inherited tendency to pressure palsies.
Specifications

Specifications

PMP22
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
Q01453
CMT1A, CMT1E, DSS, GAS-3, HMSNIA, HNPP, Sp110, PMP-22, growth arrest-specific 3, growth arrest-specific protein 3
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5376
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
PMP22
Liquid
PMP22
Synthetic peptide from near C-terminus of human PMP-22.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit