Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMPCB Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524912
Description
PMPCB is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex.Specifications
PMPCB | |
Polyclonal | |
Unconjugated | |
O75439, Q9CXT8 | |
PMPCB | |
KLH conjugated synthetic peptide between 18-46 amino acids from the N-terminal region of human PMPCB | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PMPCB | |
Beta-MPP, MPP11, MPPB, MPPP52, P-52, mitochondrial processing peptidase beta subunit, mitochondrial-processing peptidase subunit beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
73078, 9512 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction