Learn More
PNMT Guinea Pig anti-Rat, Polyclonal, Invitrogen™
Guinea Pig Polyclonal Antibody
Manufacturer:  Invitrogen PA136026
Catalog No. 01-672-216
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
For immunofluorescence applications, incubate overnight at 4°C. Store reconstituted material in small aliquots at -20°C. Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
PNMT catalyzes the conversion of norepinephrine to epinephrine in catecholamine biosynthesis.
Specifications
Specifications
PNMT
Polyclonal
whole serum with 0.1% sodium azide
P10937
PNMT
Bovine phenylethanolamine-N-methyltransferase.
50 μL
4° C
Primary
24661
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Pnmt
Lyophilized
Guinea Pig
IgG
RUO
Antibody
Polyclonal
Rat