Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNOC Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526321

Catalog No. PIPA526321

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

PNOC is the ligand of the opioid receptor-like receptor (OPRL1). It may act as a transmitter in the brain by modulating nociceptive and locomotor behavior. This protein may be involved in neuronal differentiation and development
Specifications

Specifications

PNOC
Polyclonal
Unconjugated
PNOC
Liquid
PNOC
KLH conjugated synthetic peptide between 126-151 amino acids from the central region of human PNOC
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q13519
PPNOC, nociceptin, nocistatin, propronociceptin
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5368
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit