Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNPLA8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524642

Catalog No. PIPA524642


Add to cart

Description

Description

PNPLA8 catalyzes cleavage of fatty acids from phospholipids, thereby regulating membrane physical properties and the release of lipid second messengers and growth factors. Phospholipase A2 activity also modulates cellular growth programs, inflammation, and ion channel function.
Specifications

Specifications

PNPLA8
Polyclonal
PBS with 0.09% sodium azide
Q9NP80
IPLA2(GAMMA), IPLA2-2, IPLA2G, PNPLA-gamma, calcium-independent phospholipase A2-gamma, iPLA2 gamma, iPLA2-gamma, intracellular membrane-associated calcium-independent phospholipase A2 gamma, membrane-associated calcium-independent phospholipase A2 gamma, patatin-like phospholipase domain-co
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
50640
Flow Cytometry, Western Blot
Unconjugated
PNPLA8
Liquid
PNPLA8
KLH conjugated synthetic peptide between 212-240 amino acids from the N-terminal region of human PNPLA8
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit