Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNPO Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526400

Catalog No. PIPA526400

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The enzyme encoded by this gene catalyzes the terminal, rate-limiting step in the synthesis of pyridoxal 5 ft.-phosphate, also known as vitamin B6. Vitamin B6 is a required co-factor for enzymes involved in both homocysteine metabolism and synthesis of neurotransmitters such as catecholamine.
Specifications

Specifications

PNPO
Polyclonal
PBS with 0.09% sodium azide
Q9NVS9
PDXPO, pyridoxal 5′-phosphate synthase, pyridoxamine-phosphate oxidase, pyridoxine 5′-phosphate oxidase, pyridoxine-5′-phosphate oxidase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55163
Western Blot
Unconjugated
PNPO
Liquid
PNPO
KLH conjugated synthetic peptide between 232-261 amino acids from the C-terminal region of human PNPO
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit