Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNR Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1809

Catalog No. PA1809


Add to cart

Description

Description

PA1-809 detects PNR from rat and human samples. PA1-809 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~42 kDa protein representing PNR from rat tissue extracts. The PA1-809 immunogen is a synthetic peptide corresponding to residues R(392) K T I G N T P M E K L L C D M F K N(410) of human PNR. This sequence is completely conserved between mouse and chicken. This immunizing peptide (cat. No. PEP-237) can be used for neutralization and control experiments.

The photoreceptor-specific nuclear receptor (PNR) is a recently identified nuclear transcription factor.  PNR expression pattern appears to be restricted in the retina, specific to photoreceptor cells.  PNR has been shown to share structural similarity with TLX, an orphan receptor.  Despite distinct differences in the DNA binding domain, studies have shown that PNR is capable of recognizing a subset of TLX target sequences in vitro.  Recent studies suggest that PNR plays a support role in the regulation of signaling pathways intrinsic to the photoreceptor cell function. The corresponding gene for the PNR is NR2E3.
Specifications

Specifications

PNR
Polyclonal
Unconjugated
NR2E3
Liquid
NR2E3
Synthetic peptide corresponding to residues R(392) K T I G N T P M E K L L C D M F K N(410) of human PNR.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q9Y5X4
Photoreceptor-Specific Nuclear Receptor, NR2E3
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10002, 100365683
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit