Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNUTS Mouse, Unlabeled, Clone: 47, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611060

Catalog No. BDB611060


Add to cart

Description

Description

Many aspects of cellular physiology are influenced by the serine/threonine protein phosphatase1 (PP1). It is a highly conserved eukaryotic protein that reverses the action of protein kinases. PP1 is key to such processes as cell division, neuronal metabolism, and protein synthesis. It consists of a catalytic subunit, with a broad substrate specificity, whose action is directed by its association with a family of regulatory proteins including inhibitor-1, inhibitor-2, and NIPP-1. In response to extracellular stimuli, these proteins interact with PP1 and inhibit its catalytic activity. Other regulatory proteins are referred to as targeting subunits because they direct PP1 to specific subcellular locations and modulate its activity. In the cell nucleus, PP1 has been implicated in a variety of processes. Nuclear PP1 exists in high MW complexes with other proteins including NIPP-1. PNUTS (phosphatase 1 nuclear targeting subunit) is a ubiquitously expressed nuclear protein that forms a stable complex with PP1 and is thought to modulate its catalytic activity. Thus, PNUTS is a specific regulatory protein that directs the nuclear function of PP1.

Immunofluorescence, Western Blotting

Specifications

Specifications

PNUTS
47
Unconjugated
Affinity Purified
Rat PNUTS aa. 605-716
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit