Learn More
Pokemon Rabbit anti-Chicken, Human, Marsupial, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523156
Catalog No. PIPA523156
Add to cart
Description
Description
Pokemon is a 584 amino acid protein containing 1 BTB (POZ) domain and 4 C2H2-type zinc fingers. It is involved in transcriptional regulation and influencing the switch from cellular proliferation to terminal differentiation. It is a cellular factor that preferentially associates with active chromatin and that can specifically stimulate Tat-activated HIV-1 transcription. Pokemon expression levels have been shown to predict biological behavior and clinical outcome. 
Specifications
Specifications
Pokemon
Polyclonal
whole serum with 50% glycerol and 0.025% sodium azide
O93567, O95365, O88939, Q9QZ48
FBI-1, FBI1, LRF, ZBTB7, ZNF857A, pokemon, HIV-1 1st-binding protein 1, HIV-1 inducer of short transcripts binding protein, POK erythroid myeloid ontogenic factor, POZ and Krueppel erythroid myeloid ontogenic factor, TIP21, TTF-I-interacting peptide 21, factor binding IST protein 1, factor that binds to inducer of short transcripts protein 1, leukemia/lymphoma-related factor, lymphoma related factor, zinc finger and BTB domain containing 7A, HIV-1 inducer of short transcripts binding protein, zi
Rabbit
IgG
RUO
Antibody
Polyclonal
Chicken, Human, Marsupial, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ZBTB7A
Liquid
ZBTB7A
Synthetic peptide (1 MAGGVDGPIGIPFPDHSSDI 20) of Pokemon protein.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
117107, 16969, 395411, 51341