Learn More
Poliovirus Type 1 Mouse anti-Virus, Clone: 14D2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen HYB2951502
Catalog No. 01-670-123
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
HYB 295-15-02 detects Poliovirus type 1 in virus samples. There is no cross-reactivity with poliovirus type 2 and 3. HYB 295-15-02 has been successfully used in ELISA procedures. HYB 295-15-02 immunogen is Polio virus type 1 (inactivated). NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Poliovirus is a single-stranded RNA virus that belongs to the Picornavirus family, characterized by their small RNA genome (7.2bk to 8.5 kb). Poliovirus is a enterovirus which primarily infects the gastroentestinal assiciated lymphoid tissue (GALT) and can eventually spread to the central nervous system, causing paralysis. Mortality, though very rare, is usually due to respiratory failure by paralysis of the intercostal muscles and diaphragm. There are three separate types of the virus: Type 1, Type 2 and Type 3.
Specifications
Specifications
Poliovirus Type 1
Monoclonal
1.11 mg/mL
PBS with 15mM sodium azide; pH 7.4
PV-1
Polio virus type 1 (inactivated).
Protein A
RUO
Antibody
Monoclonal
ELISA
14D2
Unconjugated
Liquid
Mouse
IgG2a, kappa
200 μL
4° C, store in dark
Primary
Virus