Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526937

Catalog No. PIPA526937

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse, non-human primate and rat based on sequence homology.

This gene encodes a DNA polymerase. DNA polymerases catalyze DNA-template-directed extension of the 3 ft.-end of a DNA strand. This particular polymerase, which is a member of the X family of DNA polymerases, likely plays a role in non-homologous end joining and other DNA repair processes.
Specifications

Specifications

POLL
Polyclonal
PBS with 0.09% sodium azide
Q9UGP5
BETAN, POLKAPPA, DNA polymerase beta-2, DNA polymerase beta-N, DNA polymerase kappa, DNA polymerase lambda
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27343
Western Blot
Unconjugated
POLL
Liquid
POLL
KLH conjugated synthetic peptide between 549-575 amino acids from the C-terminal region of human POLL
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit