Learn More
anti-POLR2C, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232319
Catalog No. NBP232319
Add to cart
Description
Description
POLR2C Polyclonal antibody specifically detects POLR2C in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
POLR2C
0.6 mg/mL
Western Blot 1:1000
P19387
DNA-directed RNA polymerase II 33 kDa polypeptide, DNA-directed RNA polymerase II subunit C, DNA-directed RNA polymerase II subunit RPB3, hRPB33, hsRPB3, polymerase (RNA) II (DNA directed) polypeptide C (33kD), polymerase (RNA) II (DNA directed) polypeptide C, 33kDa, RNA polymerase II subunit 3, RNA polymerase II subunit B3, RPB3, RPB31, RPB33
Rabbit
IgG
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
POLR2C
This POLR2C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 200-228 amino acids from the C-terminal region of human POLR2C.
31.441 kDa
400 ul
DNA Repair, DNA replication Transcription Translation and Splicing
Primary
5432