Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

POLR2L, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA125692

Catalog No. PIPA125692

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The product of this gene contains four conserved cysteines characteristic of an atypical zinc-binding domain. Like its counterpart in yeast, this subunit may be shared by the other two DNA-directed RNA polymerases.

PA1-25692 detects POLR2L from human and mouse samples.PA1-25692 has been successfully used in Western blot procedures.The PA1-25692 immunogen is full length protein.
Specifications

Specifications

POLR2L
Polyclonal
Unconjugated
POLR2L
Purified
POLR2L
Full length protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Western Blot
1mg/mL
PBS with no preservative
P62875, P62876
RBP10, RPABC5, RPB10, RPB10beta, RPB7.6, hRPB7.6, hsRPB10b
Chicken
IgY
50μg
Store at -20°C
Primary
5441
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit