Learn More
POMGNT1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA521630
Catalog No. PIPA521630
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA5-21631 targets SLC26A8 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-21631 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 613 and 850 (Q96RN1) of SLC26A8.
Specifications
Specifications
POMGNT1
Polyclonal
Unconjugated
POMGNT1
Liquid
POMGNT1
Recombit fragment corresponding to a region within amino acids 98 and 373 of POMGNT1 (Uniprot IDNo.Q8WZA1)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.9 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8WZA1
RP11-322N21.3, GNTI.2, GnT I.2, MDDGA3, MDDGB3, MDDGC3, MEB, MGAT1.2, gnT-I.2, UDP-GlcNAc:alpha-D-mannoside beta-1,2-N-acetylglucosaminyltransferase I.2, protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55624