Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PON1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals 34630002

Catalog No. 34-630-002

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PON1 Polyclonal antibody specifically detects PON1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

PON1
Unconjugated
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative
Affinity Purified
PON1
This antibody is specific for the N Terminus Region of the target protein.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This product is specific for Human PON1.
Western Blot, ELISA
Western Blot 1:5000-1:20000, ELISA 1:100-1:2000
P27169
A-esterase 1, Aromatic esterase 1, arylesterase B-type, EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, ESAesterase A, K-45, MVCD5, paraoxonase 1, PON 1, PONparaoxonase B-type, serum aryldiakylphosphatase, Serum aryldialkylphosphatase 1, serum paraoxonase/arylesterase 1
Rabbit
IgG
0.05 mg
Alzheimers Research, Cancer, Lipid and Metabolism, Neurodegeneration, Neuroscience
Primary
5444
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only