Learn More
PON2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525663
Catalog No. PIPA525663
Add to cart
Description
Description
PON2 is a member of the paraoxonase gene family, which includes three known members located adjacent to each other on the long arm of chromosome 7. The encoded protein is ubiquitously expressed in human tissues, membrane-bound, and may act as a cellular antioxidant, protecting cells from oxidative stress. Hydrolytic activity against acylhomoserine lactones, important bacterial quorum-sensing mediators, suggests the encoded protein may also play a role in defense responses to pathogenic bacteria. Mutations in this gene may be associated with vascular disease and a number of quantitative phenotypes related to diabetes.
Specifications
Specifications
PON2
Polyclonal
Unconjugated
PON2
Liquid
PON2
KLH conjugated synthetic peptide between 74-101 amino acids from the central region of human PON2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q62086, Q15165
A-esterase 2, PON 2, aromatic esterase 2, paraoxonase nirs, serum aryldialkylphosphatase 2, serum paraoxonase/arylesterase 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
330260, 5445