Learn More
Pontin (Tip49) Rabbit anti-Human, Mouse, Canine, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061299
Catalog No. 06-129-9MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use Anti-Pontin (Tip49) Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunocytochemistry to detect Pontin (Tip49) also known as 49 kDa TATA box-binding protein-interacting protein or Nuclear matrix protein 238.
Specifications
Specifications
Pontin (Tip49)
Unconjugated
Affinity Purified
Rabbit
Affinity Purified
2-8°C for 12 months
Polyclonal
Canine, Human, Murine
Immunocytochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
ECP-54; ECP54; INO80H; NMP238; PONTIN; Pontin52; RVB1; Rvb1; TAP54-alpha; TIH1; TIP49; TIP49A; TIP49a
KLH-conjugated linear peptide corresponding to human pontin.
100μg
Primary
NP_003698