Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-POP1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP126606

Catalog No. NBP126606

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

POP1 Polyclonal antibody specifically detects POP1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative), Immunocytochemistry.
Specifications

Specifications

POP1
1.0 mg/mL
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Immunoprecipitation 5-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:2000 - 1:10000, Western Blot (Negative), Immunocytochemistry
NP_055844.1
EC 3.1.26.5, hPOP1, KIAA0061, MGC17365, processing of precursor 1, ribonuclease P/MRP subunit (S. cerevisiae), processing of precursors 1, ribonucleases P/MRP protein subunit POP1
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
POP1
The immunogen recognized by this antibody maps to a region between residue 925 and 975 of human processing of precursor 1 using the numbering given in entry NP_055844.1 (GeneID 10940).
Immunogen affinity purified
RUO
Primary
10940
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only