Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IgM Isotype Control, Novus Biologicals™
Pig Antibody
Supplier: Novus Biologicals NBP196788
Description
IgM specifically detects IgM in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
IgM | |
Unconjugated | |
0.1 M Tris Chloride, 0.5 M Sodium Chloride, pH 8.0. Stabilizer: None. with 0.1% Sodium Azide | |
Porcine | |
1 mg | |
Isotype Control (Primary) | |
No reaction was observed against anti-Swine IgG F(c). Some light chain cross reactivity will occur with anti-Swine IgG. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgM |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, SDS-Page | |
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
Multi-step | |
RUO | |
3507 | |
Porcine | |
Concentrated |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction