Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141147

Catalog No. 01-675-724


Add to cart

Description

Description

Suggested positive control: HL-60, antigen standard for POT1 (transient overexpression lysate).

POT1 is a member of the telombin family and encodes a nuclear protein involved in telomere maintece. Specifically, this protein functions as a member of a multi-protein complex that binds to the TTAGGG repeats of telomeres, regulating telomere length and protecting chromosome ends from illegitimate recombination, catastrophic chromosome instability, and abnormal chromosome segregation. Increased transcriptional expression of this gene is associated with stomach carcinogenesis and its progression. Alternatively spliced transcript variants have been described.
Specifications

Specifications

POT1
Polyclonal
Unconjugated
POT1
Liquid
POT1
Synthetic peptide corresponding to residues 450-500 of human POT1.
Protein G
RUO
Antibody
Polyclonal
Human
Gel Shift, Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9NUX5
DKFZp586D211, HPOT1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
25913
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit