Learn More
MilliporeSigma™ anti-Potassium Channel HERG, C-terminus; Polyclonal; ™ (Chemicon™)
Supplier: MilliporeSigma™ AB590850UL/DEL
Description
Immunocytochemistry, Immunoprecipitation, Western Blotting
Specifications
Potassium Channel HERG, C-terminus | |
Unconjugated | |
GST fusion protein corresponding to amino acids 1106-1159 of human erg1 (HERG) (Accession number Q12809). | |
50 μL | |
Neuroscience | |
Human |
Polyclonal | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary |