Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KV1.4 (KCNA4) Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112459

Catalog No. 01-674-147


Add to cart

Description

Description

In neurons voltage-dependent potassium channels are key determits of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
Specifications

Specifications

KV1.4 (KCNA4)
Polyclonal
PBS with 0.08% sodium azide
P15385
KCHAN, Kv1.4, Kv4, RHK1, RK3
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
25469
Western Blot
Unconjugated
Kcna4
Liquid
KCNA4
Synthetic peptide.
Purified
RUO
Antibody
Polyclonal
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit