Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Potassium Channel Kv3.1, HRP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP142819H

Catalog No. NBP142819H


Add to cart

Description

Description

Potassium Channel Kv3.1 Polyclonal antibody specifically detects Potassium Channel Kv3.1 in Human, Mouse, Rat, Bovine, Canine, Rabbit samples. It is validated for Western Blot.
Specifications

Specifications

Potassium Channel Kv3.1
HRP
PBS with No Preservative
FLJ41162, FLJ42249, FLJ43491, KV3.1, KV4, MGC129855, NGK2, potassium voltage-gated channel subfamily C member 1, potassium voltage-gated channel, Shaw-related subfamily, member 1, voltage-gated potassium channel protein KV3.1, Voltage-gated potassium channel subunit Kv3.1, Voltage-gated potassium channel subunit Kv4
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Does not cross-react with Kv1.2 or Kv1.3.
Western Blot
Western Blot
Affinity Purified
KCNC1
Synthetic peptide made to an internal portion of rat Kv3.1 (within residues 350-400). [Swiss-Prot# P25122]
110 kDa
0.1 ml
Neuronal Cell Markers, Neuroscience, Neurotransmission, Potassium Channels
Primary
3746
Bovine, Canine, Human, Mouse, Rabbit, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only