Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Potassium Channel Kv3.1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$76.10 - $269.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Synthetic peptide made to an internal portion of rat Kv3.1 (within residues 350-400). [Swiss-Prot# P25122]
Antigen Potassium Channel Kv3.1
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP142819SS
View Documents
Novus Biologicals
NBP142819SS
0.025 ml
Each for $76.10
Add to cart
 
NBP142819
View Documents
Novus Biologicals
NBP142819
0.1 ml
Each for $269.50
Add to cart
 
Description

Description

Potassium Channel Kv3.1 Polyclonal antibody specifically detects Potassium Channel Kv3.1 in Human, Mouse, Rat, Bovine, Canine, Rabbit samples. It is validated for Western Blot, Immunoprecipitation, Block/Neutralize.
Specifications

Specifications

Rabbit
Synthetic peptide made to an internal portion of rat Kv3.1 (within residues 350-400). [Swiss-Prot# P25122]
Polyclonal
Primary
Western Blot, Immunoprecipitation, Blocking Assay
P25122
PBS with 0.1% Sodium Azide
IgG
Potassium Channel Kv3.1
Affinity Purified
RUO
Does not cross-react with Kv1.2 or Kv1.3.
3746
1 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit