Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KV4.2 (KCND2) Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112460

Catalog No. 01-674-148


Add to cart

Description

Description

A suggested positive control for this product is heart or brain lysate.

In neurons voltage-dependent potassium channels are key determits of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
Specifications

Specifications

KV4.2 (KCND2)
Polyclonal
PBS with 0.08% sodium azide
Q63881
Kv4.2, RK5, Shal1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
65180
Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Kcnd2
Liquid
KCND2
Synthetic peptide corresponding to aa 453-470 of rat Kv4.2 conjugated to KLH
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit