Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POTEE Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523521

Catalog No. PIPA523521


Add to cart

Specifications

Specifications

POTEE
Polyclonal
PBS with 0.09% sodium azide
Q6S8J3
A26C1, A26C1A, CT104.2, POTE-2, POTE2, POTE2gamma, ANKRD26-like family C member 1A, ANKRD26-like family C, member 1A, POTE ankyrin domain family member E, cancer/testis antigen family 104, member 2, prostate, ovary, testis-expressed protein on chromosome 2, protein expressed in prostate, ovary, testis, and pla
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
445582
Flow Cytometry, Western Blot
Unconjugated
POTEE
Liquid
POTEE
KLH conjugated synthetic peptide between 380-409 amino acids from the central region of human POTEE
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit