Learn More
Oct-2, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA120915
Catalog No. PIPA120915
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Oct2 is a transcription factor that specifically binds to the octamer motif (5'-ATTTGCAT-3'). Oct2 regulates transcription in a number of tissues in addition to activating immunoglobulin gene expression. It also modulates transcription transactivation by NR3C1, AR and PGR. Oct2 is B-cell specific, belongs to the POU transcription factor family class-2 subfamily and contains 1 homeobox domain.
PA1-20915 detects Oct2 in Human samples.PA1-20915 has been successfully used in Immunohistochemistry (Paraffin), Immunoprecipitation and Western Blot procedures.PA1-20915 immunogen corresponds to Synthetic peptide corresponding to the C-terminal region of the human protein..
Specifications
Specifications
POU2F2
Polyclonal
Unconjugated
POU2F2
Purified
POU2F2
Synthetic peptide corresponding to the C-terminal region of the human protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.2mg/mL
PBS with 1% BSA and 0.09% sodium azide; pH 7.4
P09086
OCT2, OTF2, Oct-2
Rabbit
IgG
500μL
4° C, do not freeze
Primary
5452