Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R14C Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523660

Catalog No. PIPA523660

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The degree of protein phosphorylation is regulated by a balance of protein kinase and phosphatase activities. Protein phosphatase-1 (PP1; see MIM 176875) is a signal-transducing phosphatase that influences neuronal activity, protein synthesis, metabolism, muscle contraction, and cell division. PPP1R14C is an inhibitor of PP1.
Specifications

Specifications

PPP1R14C
Polyclonal
PBS with 0.09% sodium azide
Q8TAE6, Q8R4S0
CPI17-like, KEPI, NY-BR-81, PKC-potentiated PP1 inhibitory protein, kinase C-enhanced PP1 inhibitor, kinase-enhanced PP1 inhibitor, protein phosphatase 1 regulatory subunit 14C, serologically defined breast cancer antigen NY-BR-81
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
76142, 81706
Immunohistochemistry, Western Blot
Unconjugated
PPP1R14C
Liquid
PPP1R14C
KLH conjugated synthetic peptide between 126-155 amino acids from the C-terminal region of human PP1R14C
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit