Learn More
PP2A Rabbit anti-Human, Mouse, Rat, Rabbit, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06222
Catalog No. 06-222-MI
Add to cart
Description
Description
Anti-PP2A Antibody, C subunit is an antibody against PP2A for use in immunoprecipitation and western blotting.
Specifications
Specifications
PP2A
Unconjugated
Serum
Rabbit
IgG
200μg
Primary
NM_002715.2; NP_002706
Immunoprecipitation, Western Blot
Immunoaffinity Purified immunoglobulin in 150mM Tris, pH 7.4, 0.02% sodium azide, 50% glycerol and 5mg/ml BSA
PPP2CA; PP2CA; PP2A-alpha; RP-C; PP2Ac
peptide corresponding to the C-terminal amino acid residues 296-309 of human protein phosphatase 2A
Unpurified
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Polyclonal
Human, Murine, Rabbit, Rat