Learn More
PP2A Mouse anti-Human, Mouse, Rat, Xenopus, Clone: 4G7, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05657
Catalog No. 05-657-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-PP2A Antibody, A subunit, clone 4G7 is an antibody against PP2A for use in western blotting.
Specifications
Specifications
PP2A
4G7
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
IgG1
200μg
Primary
Human, Murine, Rat, Xenopus
Western Blot
Unconjugated
NM_002716
PPP2R1A, PR65A, MGC786
Peptide corresponding to amino acids 6-19 (GDDSLYPIAVLIDE) of human Protein Phosphatase 2A, A subunit
Protein G chromatography
−20°C for 24 months.
Monoclonal