Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP5 Mouse, Unlabeled, Clone: 3, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611021

 View more versions of this product

Catalog No. BDB611021


Add to cart

Description

Description

Phosphorylation and dephosphorylation on serine and threonine residues is critical for signal transduction and the regulation of numerous cellular functions. Phosphorylation levels are modulated by protein kinases and phosphatases. The Ser/Thr phosphatase PP5 (also known as PPT) is a model protein of the fourth subfamily of the PPP-family of phosphatases which includes PP1, PP2A, and PP2B. PP5 is primarily located in the nucleus, demonstrates widespread tissue distribution, and is highly conserved among species. It consists of a C-terminal catalytic domain and an N-terminal TPR (tetratricopeptide repeat) domain which is involved in protein-protein interactions. The TPRs of PP5 mediate protein phosphatase activity. TPRs mediate the interaction of PP5 with various proteins including the hsp90-glucocorticoid receptor complex and the kinase domain of ANP guanylate cyclase receptor. PP5 is thought to be important in cell growth. For example, it functions upstream of p53 to regulate p21/WAF1/Cip1-mediated G1 growth arrest. Thus, PP5 is a nuclear protein phosphatase that is involved in signaling pathways of cell growth and arrest.

Immunofluorescence, Western Blotting

Specifications

Specifications

PP5
3
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
PPT
Affinity Purified
Rat PP5 aa. 36-238
150μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.