Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAR alpha Rabbit anti-Human, Mouse, Ovine, Porcine, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA132484

Catalog No. PA132484


Add to cart

Description

Description

PA1-32484 detects PPAR alpha from human, mouse, rat, pig and sheep samples. PA1-32484 has been successfully used in Western blot applications. Detects a band of approximately 52 kDa. The PA1-32484 immunogen is: Synthetic peptide surrounding amino acid 25 of human PPARa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Peroxisome proliferators are non-genotoxic carcinogens which are purported to exert their effect on cells through their interaction with members of the nuclear hormone receptor family, termed peroxisome proliferator activated receptors (PPARs). Nuclear h
Specifications

Specifications

PPAR alpha
Polyclonal
Unconjugated
PPARA
Liquid
PPARA
Synthetic peptide surrounding amino acid 25 of human PPARa.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Ovine, Porcine, Rat
Western Blot
0.5 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q07869, P23204, P37230
Peroxisome Proliferator Activated Receptor Alpha
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
19013, 25747, 397239, 443457, 5465
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit