Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PPAR gamma/NR1C3, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP177833

Catalog No. NBP177833

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

PPAR gamma/NR1C3
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Affinity Purified
PPARG
This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the amino terminus of human PPAR gamma 2.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This affinity purified antibody is directed against human PPAR gamma isoform 2 protein. A BLAST analysis was used to suggest reactivity with this protein from human and macaque based on 100% homology for the immunogen sequence. Cross reactivity with PPARg2 protein from dog and swine is likely due to 94% homology (15/16 identities) with the protein from these sources. Cross reactivity does occur with PPARg2 from mouse. Mouse PPARg2 shows 87% homology to the immunogen. Cross reactivity with PPARg2 homologues from other sources has not been determined. No reactivity is expected against other subtypes of PPAR.
ELISA, Western Blot
Western Blot 1:1000-1:3000, ELISA 1:4000-1:40000
P37231
CIMT1, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma
Rabbit
IgG
0.1 mg
Cancer, mTOR Pathway
Primary
5468
Human, Primate
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only