Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAR gamma Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525757

Catalog No. PIPA525757


Add to cart

Description

Description

Peroxisome proliferators are non-genotoxic carcinogens which are purported to exert their effect on cells through their interaction with members of the nuclear hormone receptor family termed peroxisome proliferator activated receptors (PPAR’s). Nuclear hormone receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate ligand. Studies indicate that PPAR’s are activated by peroxisome proliferators such as clofibric acid, nafenopin, and WY- 14,643, as well as by some fatty acids. It has also been shown that PPAR’s can induce transcription of acyl coenzyme A oxidase & cytochrome P450 (CYP450) A6 through interaction with specific response elements. The PPAR gamma 2 isoform appears to be induced very early in the differentiation of several cultured adipocyte cell lines, and has been suggested to be a domit regulator of the murine P2 (aP2) gene which encodes an intracellular lipid binding protein which is expressed only in adipose cells. PPAR gamma 2, like several other nuclear hormone receptors, heterodimerizes with RXR alpha.
Specifications

Specifications

PPAR gamma
Polyclonal
Unconjugated
PPARG
Liquid
PPARG
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PPARG
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P37231, P37238
CIMT1, GLM1, NR1C3, PPARG1, PPARG2, PPARgamma, PPAR gamma, PPAR-gamma, nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated nuclear receptor gamma variant 1, peroxisome proliferator-activated receptor gamma 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
19016, 5468
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit