Learn More
phosphopantothenoylcysteine decarboxylase, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human PPCDC protein.
Manufacturer: Abnova Corporation H00060490B01
Description
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. PPCDC (EC 4.1.1.36), one of the last enzymes in this pathway, converts phosphopantothenoylcysteine to 4-prime-phosphopantetheine (Daugherty et al., 2002 [PubMed 11923312]).[supplied by OMIM
Sequence: MEPKASCPAAAPLMERKFHVLVGVTGSVAALKLPLLVSKLLDIPGLEVAVVTTERAKHFYSPQDIPVTLYSDADEWEMWKSRSDPVLHIDLRRWADLLLVAPLDANTLGKVASGICDNLLTCVMRAWDRSKPLLFCPAMNTAMWEHPITAQQVDQLKAFGYVEIPCVAKKLVCGDEGLGAMAEVGTIVDKVKEVLFQHSGFQQSSpecifications
PPCDC | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human PPCDC protein. | |
PPCDC | |
FLJ14585/MDS018 | |
Mouse | |
50 μL | |
Yes | |
60490 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
BC014409.1 | |
PPCDC | |
PPCDC (AAH14409.1, 1 a.a. ∼ 204 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.