Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protein phosphatase, EF-hand calcium binding domain 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human PPEF1 protein.

Manufacturer:  Abnova Corporation H00005475B01

Catalog No. 89-005-997


Add to cart

Description

Description

This gene encodes a member of the serine/threonine protein phosphatase with EF-hand motif family. The protein contains a protein phosphatase catalytic domain, and at least two EF-hand calcium-binding motifs in its C terminus. Although its substrate(s) is unknown, the encoded protein has been suggested to play a role in specific sensory neuron function and/or development. This gene shares high sequence similarity with the Drosophila retinal degeneration C (rdgC) gene. Several alternatively spliced transcript variants, each encoding a distinct isoform, have been described. [provided by RefSeq

Sequence: MGCSSSSTKTRRSDTSLRAALIIQNWYRGYKARLKARQHYALTIFQSIEYADEQGQMQLSTFFSFMLENYTHIHKEELELRNQSLESEQDMRDRWDYVDSIDVPDSYNGPRLQFPLTCTDIDLLLEAFKEQQILHAHYVLEVLFETKKVLKQMPNFTHIQTSPSKEVTICGDLHGKLDDLFLIFYKNGLPSERNPYVFNGDFVDRGKNSIEILMILCVSFLVYPNDLHLNRGNHEDFMMNLRYGFTKEILHKYKLHGKRILQILEEFYAWLPIGTIVDNEILVIHGGISETTDLNLLHRVERNKMKSVLIPPTETNRDHDTDSKHNKVGVTFNAHGRIKTNGSPTEHLTEHEWEQIIDILWSDPRGKNGCFPNTCRGGGCYFGPDVTSKILNKYQLKMLIRSHECKPEGYEICHDGKVVTIFSASNYYEEGSNRGAYIKLCSGTTPRFFQYQVTKATCFQPLRQRVDTMENSAIKILRERVISRKSDLTRAFQLQDHRKSGKLSVSQWAFCMENILGLNLPWRSLSSNLVNIDQNGNVEYMSSFQNIRIEKPVQEAHSTLVETLYRYRSDLEIIFNAIDTDHSGLISVEEFRAMWKLFSSHYNVHIDDSQVNKLANIMDLNKDGSIDFNEFLKAFYVVHRYEDLMKPDVTNLG
Specifications

Specifications

protein phosphatase, EF-hand calcium binding domain 1
Unconjugated
No additive
NM_006240.2
PP7|PPEF|PPP7C
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5475
Western Blot
Mouse polyclonal antibody raised against a full-length human PPEF1 protein.
PPEF1
Serum
PPEF1
PPEF1 (NP_006231.2, 1 a.a. ~ 653 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit