Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PPEF1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132103

Catalog No. NBP132103



Description

Description

PPEF1 Polyclonal antibody specifically detects PPEF1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

PPEF1
0.12mg/mL
Western Blot 1:500-1:3000
Affinity Purified
PPEF1
Recombinant protein encompassing a sequence within the N-terminus region of human PPEF-1. The exact sequence is proprietary.
76 kDa
0.1 ml
Protein Phosphatase
Primary
5475
Human
Western Blot
Unconjugated
PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal
EC 3.1.3.16, PP7, PPEF, PPEF-1, PPP7C, PPP7CA, protein phosphatase 7, catalytic subunit, alpha isozyme, Protein phosphatase with EF calcium-binding domain, protein phosphatase, EF-hand calcium binding domain 1, protein phosphatase, serine/threonine type, with EF-hands, serine/threonine protein phosphatase 7, Serine/threonine-protein phosphatase 7, serine/threonine-protein phosphatase with EF-hands 1
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only