Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPFIA1, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127622
Description
May regulate the disassembly of focal adhesions. May localize receptor-like tyrosine phosphatases type 2A at specific sites on the plasma membrane. possibly regulating their interaction with the extracellular environment and their association with substra
PA1-27622 detects PPFIA1 from human, mouse, and rat samples.PA1-27622 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Liprin-alpha-1.Specifications
PPFIA1 | |
Polyclonal | |
PPFIA1 | |
FLJ41337, FLJ42630, FLJ43474, LIP.1, LIP1, LIPRIN, MGC26800 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
8500 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
Unconjugated | |
Q13136 | |
PPFIA1 | |
Synthetic peptide corresponding to human Liprin-alpha-1. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction