Learn More
anti-PPHLN1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232327
Catalog No. NBP232327
Add to cart
Description
Description
PPHLN1 Polyclonal antibody specifically detects PPHLN1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
PPHLN1
0.27 mg/mL
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:25
Q8NEY8
Gastric cancer antigen Ga50, HSPC232, MGC48786, periphilin 1, periphilin-1
Rabbit
IgG
Protein A purified
RUO
Primary
51535
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
PPHLN1
This PPHLN1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 45-66 amino acids from the N-terminal region of human PPHLN1.
52.737 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human